Decappers, E-Z Decappers™, WHEATON®
Toimittaja: DWK Life Sciences
W225352 W225350 W225351 W225353
WHEAW225352EA 1170 EUR
WHEAW225352 WHEAW225350 WHEAW225351 WHEAW225353
Decappers, E-Z Decappers™, WHEATON®
Korkinsulkijat ja -poistajat
These decappers are used to remove aluminium seals from bottles and vials with crimp or serum finish.
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Autoclavable