Näytepullot, Lab File®, WHEATON®
Toimittaja: DWK Life Sciences
224823 224821 224822 W224581 W224582 W224584 W224585 W224586 W224588 W224589 W224590 W224591 W224604 W224605 W224606 W224607 W224608 W224681 W224682 W225291 W225292 W225293 W224684 W224820
218-2217EA 332 EUR
218-2217 218-2215 218-2216 WHEAW224581 WHEAW224582 WHEAW224584 WHEAW224585 WHEAW224586 WHEAW224588 WHEAW224589 WHEAW224590 WHEAW224591 WHEAW224604 WHEAW224605 WHEAW224606 WHEAW224607 WHEAW224608 WHEAW224681 WHEAW224682 WHEAW225291 WHEAW225292 WHEAW225293 WHEAW224684 WHEAW224820
Näytepullot, Lab File®, WHEATON®
Näytevialit ja pienet näyteastiat
Clear vials made from low extractable borosilicate glass, amber vials made from borosilicate glass.
  • Sample vials with caps attached to vial help maintain cleanliness
  • Lab File® with partitioned trays provides an easy way to inventory samples or to store empty vials
  • Clear vials conform to ASTM E438 Type I, Class A and USP Type I requirements
  • Amber vials conform to USP Type I requirements for light transmission to protect light-sensitive products
  • 14B rubber lined black phenolic screw cap for aqueous samples