WHEATON® Diagnostic Vials
Toimittaja: DWK Life Sciences
219371 W216375 W216377 W219366 W216376 W219365
WHEA219371EA 2630 EUR
WHEA219371 WHEAW216375 WHEAW216377 WHEAW219366 WHEAW216376 WHEAW219365
WHEATON® Diagnostic Vials
Näytevialit ja pienet näyteastiat
These vials are manufactured from borosilicate glass that conforms to USP Type I requirements. The screw thread design eliminates the need for crimping tools.
  • Alternative to serum bottles and vials
  • Ideal for lyophilisation
  • Fits with I-Loc™ closure or screw cap with thin flange stopper
  • Heavy walled glass provides impact resistance