Dropping bottles, glass, Wheaton®
Toimittaja: DWK Life Sciences
W211735 W211734 W211754 W211755
WHEAW211735EA 336 EUR
WHEAW211735 WHEAW211734 WHEAW211754 WHEAW211755
Dropping bottles, glass, Wheaton®
Pullot Tippapullo
Manufactured from soda-lime glass material.
  • Available in clear or amber glass format
  • Amber coloured to protect light sensitive substanzes
  • Not autoclavable
Dropping bottles delivers stains or indicator solutions drop by drop. Complete with a glass stopper, a partial turn of the ground glass stopper seals the bottle.
Sertifikaatit: Material conforms to USP Type III requirements.