Diagnostic bottles, narrow neck, round, PET, with thread, WHEATON®
Toimittaja: DWK Life Sciences
W013973 W013974 W013975
WHEAW013973EA 0 EUR
WHEAW013973 WHEAW013974 WHEAW013975
Diagnostic bottles, narrow neck, round, PET, with thread, WHEATON®
Pullot Muovipullot
PET, clear or amber.
  • Provided non-sterile
  • Can be sterilized using EtO or gamma radiation
  • Lined Poylpropylene screw closures
Ideal for storage and shipment of liquid reagents, buffer solutions,
Tilaustiedot: Closures sold separately.
Pakkaus: Packaged in shrink wrap modules