Micro soxhlet extraction apparatus, WHEATON®
Toimittaja: DWK Life Sciences
415500 415502 415501 W416788
WHEA415500EA 684 EUR
WHEA415500 WHEA415502 WHEA415501 WHEAW416788
Micro soxhlet extraction apparatus, WHEATON®
Uuttolaitteet ja uutoshylsyt Soxhlet- ja Soxhlet-tyypin uuttolaitteet
Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Used for continuous extraction of analytes from a solid into an organic solvent.