WHEATON® Polypropylene Conical Graduated Centrifuge Tubes
Toimittaja: DWK Life Sciences
380100 380110 380220 380230 380240
WHEA380100EA 453 EUR
WHEA380100 WHEA380110 WHEA380220 WHEA380230 WHEA380240
WHEATON® Polypropylene Conical Graduated Centrifuge Tubes
Putket Sentrifugiputket
Ideal for centrifugation and general liquid handling purposes, especially in life science applications, helping eliminating the risk of cross contamination.
  • Manufactured from virgin, medical grade polypropylene
  • Fitted with leakproof, HDPE caps
  • Option of 15 or 50 ml capacity
  • Crisp black printed graduations with large white write on area
  • Supplied sterile by ETO
  • Supplied DNase/RNase-free, non-pyrogenic <0,25 EU/ml
  • Packed in bags
Order Now
TEKNISET TIEDOT
Tulpan materiaali HDPE HDPE HDPE
Environmentally Preferable
Graduation lisäykset 5 - 47,5 2 - 14,5