Screw caps for bottles, narrow neck, linerless, WHEATON®
Toimittaja: DWK Life Sciences
239510-02 239516-03 239512-02 239506-02 239506-03 239506-04 239506-06 239510 239510-03 239510-04 239512 239512-04 239516 239516-02 239516-04 239510-06 239506 239501-06 239501 239501-02 239501-03
WHEA239510-02EA 130 EUR
WHEA239510-02 WHEA239516-03 WHEA239512-02 WHEA239506-02 WHEA239506-03 WHEA239506-04 WHEA239506-06 WHEA239510 WHEA239510-03 WHEA239510-04 WHEA239512 WHEA239512-04 WHEA239516 WHEA239516-02 WHEA239516-04 WHEA239510-06 WHEA239506 WHEA239501-06 WHEA239501 WHEA239501-02 WHEA239501-03
Screw caps for bottles, narrow neck, linerless, WHEATON®
Tulpat ja korkit Pullojen sulkimet
Screw caps made of PP.
  • Use these caps with WHEATON leak resistant narrow mouth bottles
  • Available in natural, white, red, blue and green
  • Autoclavable
  • Very good chemical resistance