BOD bottles, with robotic stopper, WHEATON®
Toimittaja: DWK Life Sciences
227667
WHEA227667EA 1420 EUR
WHEA227667
BOD bottles, with robotic stopper, WHEATON®
Pullot BOD-pullot
Made of Type I borosilicate glass, black.
  • For light sensitive samples
  • With glass Robotic stopper
  • Additional opaque black cap
This is a standard 300 ml BOD bottle with black PVC plastic coating that blocks all visible light up to 800 nm. This inhibits the production of oxygen by algae. We recommend this bottle for use in marine photosynthesis projects in which you need to compare oxygen in a light and a dark bottle.