Crimp neck vials, E-Z Vial®, WHEATON®
Toimittaja: DWK Life Sciences
W225175-0303 225175 225173
WHEAW225175-0303EA 181 EUR
WHEAW225175-0303 WHEA225175 WHEA225173
Crimp neck vials, E-Z Vial®, WHEATON®
Näytevialit ja pienet näyteastiat
E-Z Vials are manufactured from Type I borosilicate glass and feature a 40% larger neck opening than standard opening vials.
  • Use with 11 mm aluminum seals
  • Limited volume insert
The larger opening improves sample accessibility while reducing auto sampler needle damage.
Pakkaus: Packaged in shrink-wrapped trays of 100 for convenience and cleanliness.