Reagent bottles, safety coated, WHEATON®
Toimittaja: DWK Life Sciences
221014 221017
WHEA221014EA 269 EUR
WHEA221014 WHEA221017
Reagent bottles, safety coated, WHEATON®
Pullot Lasipullot
Clear borosilicate glass with writing patch, Plastisol coated.
  • 33-430 screw cap attached
  • Polypropylene cap with PTFE liner
  • Autoclavable