Media bottles, graduated, WHEATON®
Toimittaja: DWK Life Sciences
W219440 219755 219817 219819 219820 219815 219759 219757 219715 219720 219760 219435 219437 219719 219717 219439
WHEAW219440EA 366 EUR
WHEAW219440 215-1695 215-1691 215-1692 215-1693 215-1690 215-1696 WHEA219757 WHEA219715 WHEA219720 WHEA219760 WHEA219435 WHEA219437 WHEA219719 WHEA219717 WHEA219439
Media bottles, graduated, WHEATON®
Manufactured from borosilicate glass which provides the best chemical resistance.
  • Special pour lip facilitates pouring
  • Black Phenolic PTFE rubber lined cap
  • Very good temperature resistance
Graduations and marking patch assists in measuring volume and identifying content. Hazardous communication label included with each case of bottles allows for content ID and safety notification. Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse.
Pakkaus: Packaged in partitioned carton toMore Product Information