Bottles, wide neck, with standard ground joints, WHEATON®
Toimittaja: DWK Life Sciences
216017 216019 216020 216021 W216015
WHEA216017EA 932 EUR
WHEA216017 WHEA216019 WHEA216020 WHEA216021 WHEAW216015
Bottles, wide neck, with standard ground joints, WHEATON®
Pullot Lasipullot
Borosilicate glass bottle and stopper.
  • Features no-drip pour lip
  • Ground glass stopper fits securely to prevent contamination
  • Easy to clean
  • Autoclavable
Wide mouth ideal for solids and viscous liquids.