Pullot, leveä suuaukko, pyöreät, kierrekorkki, WHEATON®
Toimittaja: DWK Life Sciences
209627 209628 209629 209630 209665 209666 209667 209668 209625 209550 209669 209425 209626 209427 209428 209545 209546 209547 209548 209549
215-4874EA 249 EUR
215-4874 215-4875 215-4876 215-4877 215-4878 215-4879 215-4880 WHEA209668 215-4872 215-4871 WHEA209669 215-4863 215-4873 215-4864 215-4865 215-4866 215-4867 215-4868 215-4869 215-4870
Pullot, leveä suuaukko, pyöreät, kierrekorkki, WHEATON®
Pullot Muovipullot
Ideal for a wide range of general laboratory applications, including packaging, storage, and transportation of liquids. Wide neck bottles are available in LDPE, HDPE, or PP formats.
  • LDPE bottles are flexible, impact-resistant, and chemical-resistant
  • HDPE bottles offer excellent chemical resistance and suitability for freezing at –100 °C
  • High-quality PP bottles offer exellent chemical and temperature resistance
  • Superior linerless double seal closure ensures leak free performance
  • Non-drip pour lip aids safe and clean pouring
  • Moulded tamper proofing bead