Erikoistestipaperit annostelijoissa, Whatman™
Toimittaja: Whatman products (Cytiva)
2651-500 2602-500A 2602-501A
WHAT2651-500EA 145 EUR
WHAT2651-500 700-9760 WHAT2602-501A
Erikoistestipaperit annostelijoissa, Whatman™
Testipaperit, pH-liuskat Paperiset indikaattorit
Cytiva offers a range of pH indicator and test papers to meet specific needs. These high-quality products combine ease-of-use with unsurpassed accuracy and consistency.
  • Convenient and portable for field use
Potassium iodide test paper: For detecting chlorine and other oxidising agents. In acid solution, oxidising agents react with the iodide in the test paper to liberate iodine. The paper turns blue in the presence of an oxidising agent (e.g., CI₂, Br₂, H₂O₂, HNO₂ and more).
Lead acetate test paper: Used for detecting hydrogen sulfide, this rapidMore Product Information