TCLP testing filters, Whatman™
Toimittaja: Whatman products (Cytiva)
5925-090 1810-110 1810-047 1810-090 1810-125 1810-142 1810-150 5925-142
WHAT5925-090EA 159 EUR
WHAT5925-090 WHAT1810-110 WHAT1810-047 WHAT1810-090 WHAT1810-125 WHAT1810-142 WHAT1810-150 WHAT5925-142
TCLP testing filters, Whatman™
Suodattimet Lasikuitusuodattimet
Cytiva's Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action.
Toxicity Characteristic Leaching Procedure (TCLP) is designed to predict the toxicity of hazardous waste. Industrial waste must comply with established TCLP limits to ensure that hazardous contaminants, known as landfill leachate, do not migrate into groundwater and threaten local drinking water sources.
By accurately characterizing the degree of contamination, materialMore Product Information