PYROSTAR™ ES-F MULTI Kit
Toimittaja: FUJIFILM WAKO CHEMICALS
WPEK4-20015 WPEK4-20003 WPEK4-20006 WPEK4-20125 WPEK4-20025 WPEK4-50015 WPEK4-50003 WPEK4-50006 WPEK4-50125 WPEK4-50025
WAKFWPEK4-20015EA 323 EUR
WAKFWPEK4-20015 WAKFWPEK4-20003 WAKFWPEK4-20006 WAKFWPEK4-20125 WAKFWPEK4-20025 WAKFWPEK4-50015 WAKFWPEK4-50003 WAKFWPEK4-50006 WAKFWPEK4-50125 WAKFWPEK4-50025
PYROSTAR™ ES-F MULTI Kit
Pikatestit patogeenien ja allergeenien osoittamiseen Endotoxin Tests
The PYROSTAR™ ES-F series of reagents represent our uniquely endotoxin specific line of Limulus Amebocyte Lysate products, which are formulated to be used either qualitatively as a gel-clot reagent or quantitatively as a Kinetic-Turbidimetric reagent.
  • Endotoxin-specific lysate, avoids false positive results from glucans
  • Available in multi-tests vials or single-test vials
  • Can be used as either a gel-clot or Kinetic-Turbidimetric Assay (KTA) reagent
  • (KTA) assays can be performed in tube reader or microplate reader
  • Gel-Clot lysate sensitivities range from 0,015 to 0,25 EU/ml
  • Available with matched control standard endotoxin (CSE)
  • 100 ul sample size when used with tube reader; 50 µl sample size when used with microplate reader
  • Measurement system: Toxinometer or MPR
This multi-test vial is designed to be used with the water bath, heat block or the Toxinometer®. With this configuration, the user dispenses 0,1 ml of dissolved LAL reagent into the appropriate reaction tube, then adding in 0,1 ml of the sample. This multi-test vial is ideal for a user testing a larger number of samples.
PYROSTAR™ ES-F reagents are available with a KTA quantitative range of either 0,001 EU/ml to 10 EU/ml, The KTA quantitative range is related to the Gel-Clot sensitivity.