Toxinometer® ET-7000/E Incubating Tube Reader
Toimittaja: FUJIFILM WAKO CHEMICALS
293-36061 292-36271
WAKF293-36061EA 13980 EUR
WAKF293-36061 WAKF292-36271
Toxinometer® ET-7000/E Incubating Tube Reader
Pikatestit patogeenien ja allergeenien osoittamiseen Endotoxin Tests
The Toxinometer® ET-7000 is a computer-operated kinetic incubating tube reader, designed to be exceptionally user-friendly. Our state-of-the-art expansion modules can be connected to allow for endotoxin testing in a wide range of fields and sample quantities.
  • Kinetic incubating tube reader
  • For use in kinetic-turbidimetric, kinetic-chromogenic, and gel-clot assays
  • Up to 16 samples with one module
  • Possible to connect up to 8 modules (up to 128 samples simultaneously)
  • Temperature settings at both 30 °C and 37 °C
  • Endotoxin determination in compliance with FDA and pharmacopoeial (USP/EP/JP) guidelines
  • Toximaster QC8 – State of the art software
  • BET monographs
The Toxinometer® ET-7000 can perform all LAL testing methodologies (gel clot, kinetic turbidimetric and chromogenic assays). The instrument has 16 wells for single-test configuration in order to easily detect erroneous readings or contamination associated with each well. The kinetic incubating tube reader has uniformity temperature settings of 30 °C and 37 °C.