VWR®, Cryo coder cap inserts
Toimittaja: VWR Collection
VWRI210-000031EA 23.2 EUR
VWRI210-000031 VWRI210-000032 VWRI210-000033 VWRI210-000034 VWRI210-000035 VWRI210-000036 VWRI210-000037 VWRI210-000038 VWRI210-000039 VWRI210-000040 VWRI210-000041 VWRI210-000042
VWR®, Cryo coder cap inserts
Korkin lisäkkeet
Product can be used as coding/identification of samples types stored in it.
Cryocoder/cap inserts are available in multiple colour options: Violet, white, blue, orange, lilac, red, tan, yellow, green, grey, pink, assorted (five colours).