Ultrapure water systems, arium® comfort I
Toimittaja: Sartorius
H2O-I-2-TOC-B H2O-I-2-TOC-T H2O-I-2-UV-B H2O-I-1-B H2O-I-1-T H2O-I-1-TOC-B H2O-I-1-TOC-T H2O-I-1-UV-B H2O-I-1-UV-T H2O-I-2-B H2O-I-2-T H2O-I-2-UV-T
VIVAH2O-I-2-TOC-BEA 15450 EUR
VIVAH2O-I-2-TOC-B VIVAH2O-I-2-TOC-T VIVAH2O-I-2-UV-B VIVAH2O-I-1-B VIVAH2O-I-1-T VIVAH2O-I-1-TOC-B VIVAH2O-I-1-TOC-T VIVAH2O-I-1-UV-B VIVAH2O-I-1-UV-T VIVAH2O-I-2-B VIVAH2O-I-2-T VIVAH2O-I-2-UV-T
Ultrapure water systems, arium® comfort I
Vedenpuhdistusjärjestelmät Puhtaan ja ultrapuhtaan veden järjestelmät
It is compact, environmentally friendly, reliable and easy-to-use. arium® comfort I produces ASTM Type 1 ultrapure water and Type 3 pure water. The system contains state of the art reverse osmosis technology and a unique cartridge. arium® comfort I optimizes water consumption using the integrated iJust control unit. Applied for HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC-Analysis, photometry and for buffer solutions.
  • Optional integrated TOC monitor
  • Compact design
  • Flexible display and SD card slot
  • Feed water for laboratory devices such as autoclaves