Spray bottles
Toimittaja: VITLAB
53510 53610 95286 952861
VITL53510EA 79.5 EUR
VITL53510 VITL53610 VITL95286 VITL952861
Spray bottles
Pullot Aerosolisuihkepullot
White or translucent made from PP.
  • Excellent chemical resistance
  • Excellent temperature resistance
Sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle, which can be regulated from the finest mist (nebulising) to a precise liquid jet. Range: approx. 3 to 4 metres. Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography.