Instrument trays
Toimittaja: VITLAB
72298 79890 79990 72398 72498
216-0287EA 52.7 EUR
216-0287 VITL79890 VITL79990 VITL72398 VITL72498
Instrument trays
Haihdutusmaljat/Tarjottimet/Altaat Instrumenttialustat/-astiat
Made of melamine, white.
  • Tall shape
  • Rounded corners
  • Smooth surfaces, easy to clean
Practical tray for instruments, tools, and sensitive utensils. Stable and self-supporting.
Tilaustiedot: Fitting lid made of PS, please order separately.