Bottles, wide neck, PP, round, with screw cap
Toimittaja: VITLAB
101789 101589 101689 101889 101989 102089 102189
215-2980EA 79.5 EUR
215-2980 215-2978 215-2979 215-2981 215-2982 VITL102089 VITL102189
Bottles, wide neck, PP, round, with screw cap
Pullot Muovipullot
PP, transparent.
  • Very good chemical resistance
  • Heat resistant up to +125 °C, autoclavable
  • Suitable for use with microwave ovens
Can be used anywhere in the laboratory and for sample collection. Wide opening for rapid filling and dispensing of powders, pastes and liquids.