Single channel pipettes, fixed volume, MLA D-Tipper™
Toimittaja: VISTALAB TECHNOLOGIES
1051C 1055C 1060C 1152C 1052C 1056C 1053C 1057C 1058C 1153C 1141C 1054C 1059C 1159C 1162C 1148C 1149C 1160C 1163C
VIST1051CEA 540 EUR
VIST1051C VIST1055C VIST1060C VIST1152C VIST1052C VIST1056C VIST1053C VIST1057C VIST1058C VIST1153C VIST1141C VIST1054C VIST1059C VIST1159C VIST1162C VIST1148C VIST1149C VIST1160C VIST1163C
Single channel pipettes, fixed volume, MLA D-Tipper™
Pipetit ja pipetintäyttäjät Pipetit
These superior performance MLA pipettes are ideal for clinical work.
  • Constructed with precision engineered aluminum and stainless steel
  • Light plunger forces for easy operation with thumb/ forefinger
  • Colour-coded body for easy volume identification
  • In-lab calibration adjustment
  • Single-stroke 'to deliver' operation
  • Built in tip-ejector
  • Glove guard