Stereo Microscope, Mantis PIXO
Toimittaja: VISION ENGINEERING
MPH001 MPH003 MTO003 MTO004 MTO006 MTO008 MTO010 MTO015 MTO007 MTO009 MTB200 MTB201 MTB210 MTB211 MTB212
VISIMPH001EA 5028 EUR
VISIMPH001 VISIMPH003 VISIMTO003 VISIMTO004 VISIMTO006 VISIMTO008 VISIMTO010 VISIMTO015 VISIMTO007 VISIMTO009 VISIMTB200 VISIMTB201 VISIMTB210 VISIMTB211 VISIMTB212
Stereo Microscope, Mantis PIXO
Mikroskoopit
PIXO is a eyepiece-less optical stereo microscope with integrated high-definition digital camera and dynamic view optics.
  • High quality optical stereo image
  • Superb large image – no eyepieces
  • Superior ergonomics for fatigue-free viewing
  • Ergonomic working position with long working distance
  • Video and image capture
  • On screen annotation
  • Custom overlays
  • Three lens multi-view turret
Mantis is designed for precision engineering, electronic engineering, medical device manufacture, and a wide range of other applications that require high-quality images andMore Product Information