Stereo Microscope, Mantis ERGO
Toimittaja: VISION ENGINEERING
MRH001 MRH002
VISIMRH001EA 3474 EUR
VISIMRH001 VISIMRH002
Stereo Microscope, Mantis ERGO
Mikroskoopit
ERGO eyepiece-less optical stereo microscope with dynamic view optics.
  • High quality optical stereo image
  • Ergonomic working position
  • Long working distance
  • Magnification 3× to 15×
  • 3 lens multi-view turret
  • Dynamic 3D lighting
  • White/UV lighting option
Mantis is designed for precision engineering, electronic engineering, medical device manufacture, and a wide range of other applications that require high-quality images and superior ergonomics. It features a unique, patented, eyepiece-less design that delivers a large, high-quality optical stereo imageMore Product Information