Stereo microscopes, Lynx EVO
Toimittaja: VISION ENGINEERING
EVO502 EVO501 EVL045 EVL062 EVL150 EVL200 EVL100
VISIEVO502EA 11683 EUR
VISIEVO502 VISIEVO501 VISIEVL045 VISIEVL062 VISIEVL150 VISIEVL200 VISIEVL100
Stereo microscopes, Lynx EVO
Mikroskoopit
High productivity eyepiece-less stereo microscope.
  • Unrivalled ergonomics improves productivity
  • Advanced optics allows fast and accurate inspection
  • Flexibility for a wide range of applications
  • Lynx EVO stand options (Ergo stand or multi-axis stand)
Lynx EVO is a high productivity stereo microscope without eyepieces, powering your productivity through stunning 3D imaging. The unique patented eyepiece-less optics of Lynx EVO liberates users from restrictive working practices, opening up a world of enhanced efficiency through unrivalled ease of use andMore Product Information