Versilon™ C-544-A IB Tubings
Toimittaja: Saint Gobain Life Sciences
AZY02019 AZY02029 AZY02064
VERNAZY02019EA 673 EUR
VERNAZY02019 VERNAZY02029 VERNAZY02064
Versilon™ C-544-A IB Tubings
Letku Siirtoputki
Versilon™ C-544-A IB flexible, reinforced tubing from Saint-Gobain is designed for demanding applications requiring a tight bend radius where other tubes have collapsed and failed. Specially formulated from tough ether-based polyurethane resins, this tubing also retains much of its flexibility even at temperatures as low as −73 °C (−100 °F).
  • Exceptional abrasion and tear resistance
  • Tough braid reinforcement for elevated working pressures
  • Excellent resistance to oils, greases and fuels
  • Retains flexibility in sub-zero environments
Reinforced forMore Product Information