Laboratory tubing, Tygon® LMT-55
Toimittaja: Saint Gobain Life Sciences
ALH00017 ALH00018 ALH00022 ALH00023 ALH00005 ALH00007 ALH00012 ALH00013 ALH00027 ALH00002
VERNALH00017EA 135 EUR
VERNALH00017 VERNALH00018 VERNALH00022 VERNALH00023 VERNALH00005 VERNALH00007 VERNALH00012 VERNALH00013 VERNALH00027 VERNALH00002
Laboratory tubing, Tygon® LMT-55
Letku Siirtoputki
Tygon®, non-DEHP, crystal clear.
  • Outstanding chemical resistance
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Lot-to-lot consistency for reproducible results
  • Increases productivity in peristaltic pumps
  • Non-oxidising and non-contaminating
  • Smooth, polished inner wall
  • Contains no BPA
Tubing handles most inorganic chemicals in the laboratory. It is non oxidising and non contaminating, and less permeable than rubber tubing. The glassy, smooth inner bore helps prevent build up to facilitate cleaning, andMore Product Information