High-purity tubing, Tygon® 2475
Toimittaja: Saint Gobain Life Sciences
ACG00003 ACG00007 ACG00012 ACG00017 ACG00022 ACG00027 ACG00038 ACG00046 ACG00053 ACG42064
VERNACG00003EA 178 EUR
VERNACG00003 VERNACG00007 VERNACG00012 VERNACG00017 VERNACG00022 VERNACG00027 VERNACG00038 VERNACG00046 VERNACG00053 VERNACG42064
High-purity tubing, Tygon® 2475
Letku Siirtoputki
Tygon®, High-purity tubing providing a high level of purity not previously available in a clear, flexible tubing.
  • Exceptionally low absorption and adsorption compared to silicone
  • Smooth inner surface inhibits particulate entrapment
  • Environmentally safer to dispose
  • Highly resistant to aggressive cleaners and sanitisers
  • Plasticiser free
Applications includes sterile filling and dispensing systems, diagnostic and nuclear equipment, laboratory analytical instrumentation, infusion sets for parenterals and drugs, cosmetic production, food and beverageMore Product Information