Digital Homogenisers, OV 725
Toimittaja: VELP SCIENTIFIC
F20900475 SA20900475
VELPF20900475EA 2120 EUR
VELPF20900475 VELPSA20900475
Digital Homogenisers, OV 725
Homogenisaattorit Homogenisaattorit, pöytämalli
Premium digital homogeniser with advanced safety featuring a powerful brushless motor, electronic speed control, integral timer and counter function. Engineered for homogenisation, emulsification, suspensions, and high-speed mixing of cosmetic, food, and pharmaceutical products, as well as biological tissues.
  • Powerful brushless motor running from 2000 up to 25000 rpm
  • Volume ranges from 0,2 ml to 2,5 L (H₂O); handles samples up to 5000 mPa*s viscosity
  • Control panel is designed to optimise user experience with the bright and intuitive 4-digit LED display