Automatic solvent extractors, SER 158 series
Toimittaja: VELP SCIENTIFIC
S303A0390 S303A0380 F303A0390 F303A0380
VELPS303A0390EA 0 EUR
VELPS303A0390 VELPS303A0380 VELPF303A0390 VELPF303A0380
Automatic solvent extractors, SER 158 series
Uuttolaitteet ja uutoshylsyt Automaattiset uutinjärjestelmät
A fully automated and cloud enabled solvent extractor capable of a high sample throughput, offering state-of-the-art technology for a fast, precise and accurate extractions in complete safety according to Randall or Twisselmann techniques.
  • Exceptional versatility and scalability
  • High productivity and performance
  • Extreme safety
  • Smart user interface
  • Cloud-enabled