Digital Homogenisers, OV 625
Toimittaja: VELP SCIENTIFIC
F20900470 SA20900470
VELPF20900470EA 1380 EUR
VELPF20900470 181-0675
Digital Homogenisers, OV 625
Homogenisaattorit Homogenisaattorit, pöytämalli
Powerful digital homogeniser designed for the maximum level of reproducibility in a wide range of applications. Cutting-edge technology and engineering expertise guarantee precision and high shearing force for fast dispersion, homogenisation, emulsification, suspensions, and high-speed mixing of biological tissues, pharmaceuticals, cosmetics and food products.
  • Volume range from 0,2 ml to 2,5 L (H₂O), handles samples up to 5000 mPas viscosity
  • Bright 3-digit LED display and the comfortable rotating knob enable precise electronic speed settings from 3000 to 25