Chemistry vacuum pumps, ME 16 NT and ME 16 NT + EK
Toimittaja: VACUUBRAND
20741500 741302 20741300
188-0005EA 12340 EUR
188-0005 VACU741302 VACU20741300
Chemistry vacuum pumps, ME 16 NT and ME 16 NT + EK
Pumput Membraanipumput
Chemistry diaphragm pumps of the NT design are an excellent solution for continuous, oil-free pumping of corrosive gases and vapors. The one-stage construction provides the advantageous combination of high pumping speed and low ultimate vacuum down to 70 mbar. All major parts in contact with pumped media are made of chemically resistant fluoroplastics. Well-proven PTFE sandwich diaphragms for increased reliability and extended operating life. The eight cylinders pump ME 16C NT provides a particularly high performance together with a compact design. Upgraded with an outlet exhaust vaporMore Product Information