High visibility bump caps, uvex u-cap sport hi-viz
Toimittaja: UVEX
9794.491 9794.490 9794.480 9794.481 9794.482 9794.492
UVEX9794.491EA 34.3 EUR
UVEX9794.491 UVEX9794.490 UVEX9794.480 UVEX9794.481 UVEX9794.482 UVEX9794.492
High visibility bump caps, uvex u-cap sport hi-viz
Kypärät ja kolhulippikset Bump Caps
These caps in fluorescent yellow or fluorescent orange feature an innovative hard shell element for reliable impact protection.
  • Protect against abrasions and impact injuries
  • Can be quickly and easily fitted to the head using the adjustable velcro fastening
  • Short peak for a wider field of upward vision
  • Perfect addition to high visibility clothing
Sertifikaatit: EN 471, EN 812