Safety welding goggles, Ultrasonic Flip-up
Toimittaja: UVEX
9302.045 9302.043
UVEX9302.045EA 62.8 EUR
UVEX9302.045 UVEX9302.043
Safety welding goggles, Ultrasonic Flip-up
Suojalasit, naamiomalliset
Wide-vision welding safety goggles feature a sporty design, and are also suitable as overspecs.
  • Perfect colour recognition thanks to Uvex sunglare filters
  • New filter technology combines UV 400 and IR protection
  • Filter extremely scratch-resistant on both sides, minimizing burns due to welding sparks (uvex infradur)
  • No problems with pressure when worn for long periods, due to a combination of hard and soft components
The combination of hard and soft material components used in the uvex ultrasonic pairs high mechanical strength with optimum wearability.More Product Information