Cover spectacles, uvex mercury, 9155
Toimittaja: UVEX
9155.005
UVEX9155.005EA 96.5 EUR
UVEX9155.005
Cover spectacles, uvex mercury, 9155
Suojalasit
Slim men’s dual lens glasses provides maximum wearer comfort.
  • Spectacles fit well in breast pocket due to flat design
  • Transparent side shields for increased protection
  • Excellent peripheral vision
Sertifikaatit: EN 166, EN 170