Anti-TRIM35 Goat Polyclonal Antibody

Toimittaja: US Biological
T8466-35C
USBIT8466-35CEA 640 EUR
USBIT8466-35C
Anti-TRIM35 Goat Polyclonal Antibody
Vasta-aineet
Recommended dilution: ELISA: 1:32000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: TRIM35 (tripartite motif containing 35)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR