Anti-TLR2 Mouse Monoclonal Antibody (Biotin) [clone: 9G273]

Toimittaja: US Biological
T8050-09Q
USBIT8050-09QEA 539 EUR
USBIT8050-09Q
Anti-TLR2 Mouse Monoclonal Antibody (Biotin) [clone: 9G273]
Vasta-aineet
Recommended dilution: Immunochemistry: 1:50; Flow Cytometry: 1:50

Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: 9G273
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR