Anti-TASP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
T1023-90A-PE
USBIT1023-90A-PEEA 884 EUR
USBIT1023-90A-PE
Anti-TASP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:250-1:500; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: TASP1 (taspase, threonine aspartase, 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR