Anti-Shigella sp. Rabbit Polyclonal Antibody

Toimittaja: US Biological
S1013-15E
USBIS1013-15EEA 534 EUR
USBIS1013-15E
Anti-Shigella sp. Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: Shigella sp.
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR