Anti-SCAP Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
S0391-10-PE
USBIS0391-10-PEEA 884 EUR
USBIS0391-10-PE
Anti-SCAP Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: SCAP (SREBF chaperone)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR