Anti-Nucleoside Diphosphate Kinase D Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
N7100-63C-PE
USBIN7100-63C-PEEA 884 EUR
USBIN7100-63C-PE
Anti-Nucleoside Diphosphate Kinase D Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: Nucleoside Diphosphate Kinase D
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR