Anti-Nuclear Receptor Subfamily 2 Group C Member 2 Mouse Monoclonal Antibody [clone: 9G407]

Toimittaja: US Biological
N6889-94D
USBIN6889-94DEA 601 EUR
USBIN6889-94D
Anti-Nuclear Receptor Subfamily 2 Group C Member 2 Mouse Monoclonal Antibody [clone: 9G407]
Vasta-aineet
Type: Primary
Antigen: Nuclear Receptor Subfamily 2 Group C Member 2
Clonality: monoclonal
Clone: 9G407
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR