Anti-Neuropeptide B-29 Rabbit Polyclonal Antibody (Carboxyfluorescein)

Toimittaja: US Biological
N2175-91A2
USBIN2175-91A2EA 1160 EUR
USBIN2175-91A2
Anti-Neuropeptide B-29 Rabbit Polyclonal Antibody (Carboxyfluorescein)
Vasta-aineet
Type: Primary
Antigen: Neuropeptide B-29
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR