Anti-NCK1-D3, Recombinant, Human Rabbit Monoclonal Antibody [clone: 7H42]

Toimittaja: US Biological
N0548-02A
USBIN0548-02AEA 607 EUR
USBIN0548-02A
Anti-NCK1-D3, Recombinant, Human Rabbit Monoclonal Antibody [clone: 7H42]
Vasta-aineet
Type: Primary
Antigen: NCK1-D3, Recombinant, Human
Clonality: Monoclonal
Clone: 7H42
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR