Anti-MSTN Rabbit Polyclonal Antibody (FITC (Fluorescein))

Toimittaja: US Biological
M9875-01A-FITC
USBIM9875-01A-FITCEA 884 EUR
USBIM9875-01A-FITC
Anti-MSTN Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50
Do not freeze!

Type: Primary
Antigen: MSTN (myostatin)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR