Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F103]

Toimittaja: US Biological
M3887-43A
USBIM3887-43AEA 652 EUR
USBIM3887-43A
Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F103]
Vasta-aineet
Recommended dilution: Flow Cytometry:1:5;

Type: Primary
Antigen: MHC Class 2, DQ, DR, Polymorphic
Clonality: monoclonal
Clone: 10F103
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR