Anti-MHC Class 1, H2, KK Mouse Monoclonal Antibody (Biotin) [clone: 36-7-5]

Toimittaja: US Biological
M3885-85H
USBIM3885-85HEA 459 EUR
USBIM3885-85H
Anti-MHC Class 1, H2, KK Mouse Monoclonal Antibody (Biotin) [clone: 36-7-5]
Vasta-aineet
Type: Primary
Antigen: MHC Class 1, H2, KK
Clonality: monoclonal
Clone: 36-7-5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR