Anti-MAP1LC3C Goat Polyclonal Antibody

Toimittaja: US Biological
M2346-30C
USBIM2346-30CEA 568 EUR
USBIM2346-30C
Anti-MAP1LC3C Goat Polyclonal Antibody
Vasta-aineet
Recommended dilution: Immunochemistry: 3. 75-7. 5 ?g/ml

Type: Primary
Antigen: MAP1LC3C (microtubule-associated protein 1 light chain 3 gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR